MagnaNeutral™ Transfer Membrane, GVS
Supplier: GVS FILTER TECHNOLOGY
1213364 1213405 1213410
490007-456EA 710.46 USD
490007-456 490007-458 490007-460 490007-462
MagnaNeutral™ Transfer Membrane, GVS
Transfer Membranes
Neutral nylon transfer membrane.
  • Naturally hydrophilic
  • Optimized for protein binding and for highly reproducible binding of nucleic acids
  • High binding capacity
  • High strength and durability, preventing distortion or contamination in multiple re-probing
Magna Neutral Nylon Transfer Membranes are pure polymer impregnated by an inert polyester web.