Anti-Neurofilaments Mouse Monoclonal Antibody [clone: 2F11]

Supplier: Diagnostic Biosystems

PDM012 MOB080-05 MOB080
99990-836EA 287.42 USD
99990-836 95024-702 99990-264
Anti-Neurofilaments Mouse Monoclonal Antibody [clone: 2F11]
Antibodies
This antibody reacts with 70 and 200 kDa component of the neurofilament polypeptide subunit. It stains neurons, neuronal processes and peripheral nerves. This antibody reacts with some neuroblastomas and gangliomas but not with other non-neuronal tumors.

Type:
Antigen: Neurofilaments
Clonality: Monoclonal
Clone: 2F11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR