Anti-MUM1 Mouse Monoclonal Antibody [clone: MUM1p]

Supplier: Diagnostic Biosystems

MOB420 MOB420-05
99990-776EA 1303.33 USD
99990-776 95024-692
Anti-MUM1 Mouse Monoclonal Antibody [clone: MUM1p]
Antibodies
Multiple myeloma oncogene-1 (MUM1) is a 50kDa protein encoded by the MUM1 gene. This antibody stains MUM1 protein, which is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. This antibody is useful in the sub classification of lymphoid malignancies.

Type:
Antigen: MUM1
Clonality: Monoclonal
Clone: MUM1p
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR