Anti-MSH6 Mouse Monoclonal Antibody [clone: 44]

Supplier: Diagnostic Biosystems

PDM147 MOB429-05 MOB429
95029-768EA 347.01 USD
95029-768 95024-688 99990-792
Anti-MSH6 Mouse Monoclonal Antibody [clone: 44]
Antibodies
MSH6 is a heterodimer of MSH2 and binds to DNA containing G/T mismatches. The MSH2-MSH6 complex recognizes a single-base mispair insertion/deletion loop. An alteration of microsatellite repeats is the result of slippage owing to strand misalignment during DNA replication and is referred to as microsatellite instability (MSI). These defects in DNA repair pathways have been related to human carcinogenesis. Studies have shown the mutations of MLH-1, MSH2 and MSH6 genes contribute to the development of sporadic colorectal carcinoma. The repair of mismatch DNA is essential to maintaining the integrity of genetic information over time.

Type:
Antigen: MSH6
Clonality: Monoclonal
Clone: 44
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR