Anti-TIMP2 Mouse Monoclonal Antibody [clone: 3A4]

Supplier: Diagnostic Biosystems

MOB318-05 MOB318
95024-848EA 572.2 USD
95024-848 99990-596
Anti-TIMP2 Mouse Monoclonal Antibody [clone: 3A4]
Antibodies
This antibody is specific to 21 kDa protein known as TIMP-2. TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family. TIMP are of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (MMP-2) and active form of 92 kDa type IV collagenase (MMP-9). TIMP inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Type:
Antigen: TIMP2
Clonality: Monoclonal
Clone: 3A4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR