Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]

Supplier: Diagnostic Biosystems

MOB349 PDM134 MOB349-05
99990-652EA 1197.37 USD
99990-652 99991-040 95024-674
Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]
Antibodies
This antibody reacts with an unknown antigen on microvilli of mesothelioma cells. It stains normal mesothelial cells as well as epithelial mesotheliomas in a thick membrane pattern due to abundant lung microvilli on the surface of these cells.

Type:
Antigen: Mesothelioma
Clonality: Monoclonal
Clone: HBME-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR