Anti-Mart-1/ Melan-A Mouse Monoclonal Antibody [clone: A103]

Supplier: Diagnostic Biosystems

MOB277 MOB277-05 PDM153
99990-524EA 941.74 USD
99990-524 95024-656 10064-970
Anti-Mart-1/ Melan-A Mouse Monoclonal Antibody [clone: A103]
Antibodies
This antibody is specific to a protein of 20 kDa known as MART-1 (melanoma antigen recognized by T cells-1) or Melan-A. This antibody does not cross-react with MAGE-1or tyrosinase. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. This antibody stains melanomas and other tumors showing melanocytic differentiation.

Type:
Antigen: Mart-1/ Melan-A
Clonality: Monoclonal
Clone: A103
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR