Anti-Pneumocystis carinii Mouse Monoclonal Antibody [clone: 3F6]

Supplier: Diagnostic Biosystems

MOB091-05 MOB091
95024-792EA 430.47 USD
95024-792 99990-282
Anti-Pneumocystis carinii Mouse Monoclonal Antibody [clone: 3F6]
Antibodies
This antibody reacts with P. carinii in the fixed human tissues. The antibody is specific against an 82 kDa parasite specific polypeptide. It does not cross-react with G. lamblia, T. gondii, T. cruzi, L. tropica, E. histolytica, C. albicans and P. falciparum.

Type:
Antigen: Pneumocystis carinii
Clonality: Monoclonal
Clone: 3F6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR