Anti-p80 Anaplastic Lymphoma Kinase Mouse Monoclonal Antibody [clone: 5A4]

Supplier: Diagnostic Biosystems

MOB416-05 MOB416
95024-772EA 576.15 USD
95024-772 99990-768
Anti-p80 Anaplastic Lymphoma Kinase Mouse Monoclonal Antibody [clone: 5A4]
Antibodies
Anaplastic large cell lymphoma (ALCL) is usually composed of large pleomorphic cells, which express CD30 antigen and the epithelial membrane antigen (EMA). These tumor cells tend to occur in younger patients and may be associated with cutaneous and extranodal involvement. Large cell lymphoma account for approximately 25% of all non-Hodgkin lymphomas in children and young adults.

Type:
Antigen: p80 Anaplastic Lymphoma Kinase
Clonality: Monoclonal
Clone: 5A4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR