Anti-P57KIP2 Mouse Monoclonal Antibody [clone: 57P06]

Supplier: Diagnostic Biosystems

MOB291 MOB291-05
99990-550EA 925.83 USD
99990-550 95024-744
Anti-P57KIP2 Mouse Monoclonal Antibody [clone: 57P06]
Antibodies
This antibody is specific to a protein of 57 kDa known as p57Kip2 a cell cycle regulatory mitotic inhibitor. This antibody does not cross-react with p27Kip1. p57Kip2 is a potent tight-binding inhibitor of several G1 cyclin complexes and a negative regulator of cell proliferation.

Type:
Antigen: P57KIP2
Clonality: Monoclonal
Clone: 57P06
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR