Anti-P53 Mouse Monoclonal Antibody [clone: DO7]

Supplier: Diagnostic Biosystems

MOB082-05 MOB082 PDM013
95024-762EA 455.64 USD
95024-762 99990-268 99990-838
Anti-P53 Mouse Monoclonal Antibody [clone: DO7]
Antibodies
This antibody reacts with wild as well as mutant types of P53 protein. It recognizes an epitope in the N-terminus of p53 protein, known to reside between amino acids 35 to 45. This antibody can be demonstrated in 22-76% of the colon, stomach, bladder, breast, lung and testes cancers.

Type:
Antigen: P53
Clonality: Monoclonal
Clone: DO7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR