Anti-Insulin Mouse Monoclonal Antibody [clone: K36aC10]

Supplier: Diagnostic Biosystems

MOB234 MOB234-05
99990-460EA 502.01 USD
99990-460 10064-910
Anti-Insulin Mouse Monoclonal Antibody [clone: K36aC10]
Antibodies
This antibody recognizes purified insulin from the pancreas of human, as well as proinsulin from human.

Type:
Antigen: Insulin
Clonality: Monoclonal
Clone: K36aC10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR