Anti-HHV8 Mouse Monoclonal Antibody [clone: LN53]

Supplier: Diagnostic Biosystems

MOB395-05 MOB395
95024-588EA 410.6 USD
95024-588 99990-732
Anti-HHV8 Mouse Monoclonal Antibody [clone: LN53]
Antibodies
This antibody stains with ORF-73 of human herpesvirus 8 (HHV8). HHV8 is the likely etiological agent of Kaposi sarcoma (KS). HHV8 encodes a latent nuclear antigen (LNA), which is the product of the viral gene ORF 73. LNA is capable of forming a complex with retinoblastoma susceptibility gene product, which may be related to its oncogenic activity.

Type:
Antigen: HHV8
Clonality: Monoclonal
Clone: LN53
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR