Anti-Gastrin Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP044-05 RP044 PDR027
95025-030EA 316.57 USD
95025-030 99991-276 99991-094
Anti-Gastrin Polyclonal Antibody
Antibodies
The antibody is specific against gastrin-containing cells. This anti-body reacts with human gastrin I and human gastrin I fragments. No cross-reactivity was observed with rat gastrin, human big gastrin, human gastrin releasing peptide, sulfated cholecystokinin amide, non-sulfated cholecystokinin amide and cholecystokinin.
Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now