Anti-CK15 Mouse Monoclonal Antibody [clone: LKH15]

Supplier: Diagnostic Biosystems

MOB305 MOB305-05
99990-570EA 524.51 USD
99990-570 95024-474
Anti-CK15 Mouse Monoclonal Antibody [clone: LKH15]
Antibodies
Cytokeratin 15 is a type I cytokeratin without a defined type II partner. Cytokeratin 15 is expressed primarily in the basal keratinocytes of stratified tissues, including the fetal epidermis and fetal nail.

Type:
Antigen: CK15
Clonality: Monoclonal
Clone: LKH15
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR