Anti-Cytokeratin (HMW) Mouse Monoclonal Antibody [clone: 34[beta]E12]

Supplier: Diagnostic Biosystems

MOB059 MOB059-05 PDM074
99990-230EA 596.06 USD
99990-230 95024-492 99990-932
Anti-Cytokeratin (HMW) Mouse Monoclonal Antibody [clone: 34[beta]E12]
Antibodies
This antibody recognizes keratin polypeptides of 68, 58, 56.5 and 50 kDa in the stratum corneum. The antibody reacts with squamous, ductal and other complex epithelia. It stains adenocarcinomas, breast, pancreas, bile duct, salivary gland and transitional cell carcinomas.

Type:
Antigen: Cytokeratin (HMW)
Clonality: Monoclonal
Clone: 34[beta]E12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR