Variable Size Geological Thin Section Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75941-16 75941-16H
102681-252EA 390.42 USD
102681-252 102681-254
Variable Size Geological Thin Section Holder, Electron Microscopy Sciences
Slide Holders
This thin section holder is made from vacuum grade aluminum with copper clips and brass screws.
The unit holds either standard thin sections of 47×27 mm or smaller or larger thin sections. Smallest size would be 40×20 mm. The holder features two clips to insure nothing moves. The holder has convenient tweezer inserts which allow for easy loading.
Información de suministro: Supplied complete with two clips.