Dual Geological Thin Section Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75941-18 75941-18H
102681-256EA 465.47 USD
102681-256 102681-258
Dual Geological Thin Section Holder, Electron Microscopy Sciences
Slide Holders
This thin section holder made from aluminum with copper clips and brass screws.
The unit measures 55 x 58 x 8 mm and it holds two standard geological thin sections or slides of 47 x 27 mm. The holder has convenient tweezer inserts which allow for easy loading. Clips are provided for secure holding.