Metallurgical Mount Holder, 1-¹/₂" and 40 mm, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75941-08 75941-08H
102681-240EA 112.71 USD
102681-240 102681-242
Metallurgical Mount Holder, 1-¹/₂" and 40 mm, Electron Microscopy Sciences
Specimen Mounting Supplies
This set screw vice which is made from machined aluminum with stainless steel allen set screws has an open slot that is 10mm (.394”) wide x 5mm (.197”) deep. It measures 25mm (1”) dia. x 17.5mm (0.69”) high.