XTEM TEM Sample Preparation Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50110-XTEM 55110-AT1 55110-MW
102092-716EA 16132.64 USD
102092-716 102092-792 102092-802
XTEM TEM Sample Preparation Kit, Electron Microscopy Sciences
Stains Microscopy and staining reagents
XTEM TEM Sample Preparation Kit is specifically for the preparation of cross-sectional TEM (XTEM) specimens
For the study of interfaces by transmission electron microscope (TEM) it is critical to use TEM samples of ultimate quality. The kit offers a complete technology and product range for preparing cross-sectional samples of these quality including specially designed mechanical sample preparation tools and embedding ring.
Información de suministro: The kit includes Microsaw sectioning tool, Micropol grinder and polisher, Microheat embedding tool, Embedding rings (“Ti disc”) 5 of each size, Special purpose mechanical tools (HSS) for embedding, Ion beam resistant glue, LEIT-C conductive carbon cement, Thermoplastic transparent glue and Diamond paste.