Multi Holder for 49 Pin Stubs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75952-49 75952-49H
102098-318EA 685.26 USD
102098-318 102098-320
Multi Holder for 49 Pin Stubs, Electron Microscopy Sciences
Specimen Mounting Supplies
This 117 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with 302 stainless steel springs that hold the pin stubs in place.
It accommodates up to forty nine standard 12.7 mm pin stubs.