Standard Reticles with Single Cross Line, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60250-19 60250-21 60251-19 60251-21 60252-19 60252-21
102093-158EA 297.34 USD
102093-158 102093-160 102093-162 102093-164 102093-166 102093-168
Standard Reticles with Single Cross Line, Electron Microscopy Sciences
Graticules, Reticles and Grids
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.