Alum celestine blue stain for histology
Supplier: Electron Microscopy Sciences
Synonyms: Iron alum - celestine blue
26023-02 26023-05
102092-366EA 71.31 USD
102092-366 102092-368
Alum celestine blue stain for histology
Alum celestine blue
A stain solution for nuclei. A substitute for Hematoxylin/Eosin.
  • High quality staining solution
  • Ready-to-use
Lendrum & McFarlane, (1940). J. of Pathology & Bacteriology, v. 50, pp 381