CMCP Macroinvertebrate, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Mounting Medium: CMCP is a colorless, non resinous, water miscible mounting medium for permanent transparent mounts
This medium used like Canada Balsam, it can be used to mount almost any material, provided it is not stained, calciferous material, or thin tissue sections. This mounting media has the advantage of allowing live or preserved specimens to be mounted directly from water or alcohol. Free living nematodes and live parasitic worms are among the organism suitable for mounting with CMCP. The CMCP mounting provide then with rapid relaxation, death, and clearing action.