SEM Single Mount Storage Tube and Mailer, Hitachi, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76531-05 76531-10 76535-01
102098-416PK 99.53 USD
102098-416 102098-418 102098-420
SEM Single Mount Storage Tube and Mailer, Hitachi, Electron Microscopy Sciences
Mailers and Shippers
Ideal for storage as well as mailing.
A 15 mm diamter plastic tube and plug is for M4 threaded Hitachi mounts. A 15 mm Hitachi mount can be inserted securely to the plug and then protected by the tube. The internal diameter of the tube is 21 mm (0.83”). The Outer measurements of the tube are: 23 x 52 mm (0.9 x 2.05").