Reticles with Frame, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67960 67961 67962
100496-238EA 88.79 USD
100496-238 100496-240 102091-108
Reticles with Frame, Electron Microscopy Sciences
Graticules, Reticles and Grids
Reticle A is designed for general laboratory use (5 mm long, with 0.1 mm divisions) and reticle B (4/16 " long, 1/64" divisions) is most frequently used in textiles or electronics, and where measurement in mils is required. Reticle C (5000 µm long, with 50 µm divisions) is calibrated in microns and helpful for pesticide droplet size measurements or anywhere measurements in microns are required.