Flexible Mailers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61512-70 61512-75
102093-424PK 83.31 USD
102093-424 102093-426
Flexible Mailers, Electron Microscopy Sciences
Mailers and Shippers
Includes all required secondary and outer packaging components. Users provide only leak-proof specimen containers (primary receptacles), such as tubes, vials, cups, etc. Accommodates any size and quantity of specimen containers that fit inside the flexible secondary envelope with required absorbent material.
  • Maximum capacity is 300 ml or less
Certificaciones: Complies fully with US postal service, DOT and IATA regulations for transport of clinical and diagnostic specimens (UN3373) by air or ground; Domestic Mail Manual Part 8.6; Title 49 CFR, Part 173, 199; and IATA Packing Instruction 650.