MicroTool Kits, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60100-01 60100-02 60100-03
102092-864EA 268.53 USD
102092-864 102092-866 102092-868
MicroTool Kits, Electron Microscopy Sciences
Specimen Mounting Supplies
General use: for protein crystals, single cells and manipulating small samples.
These patent-pending MicroTools™ have tips made from soft, flexible microfabricated polymer films. The curvature of the tips gives them rigidity, but they can still easily be flexed. Tools designed for use with samples from a few micrometers to millimeter in size. These tools will cause far less  damage to fragile samples than metal tools, and they are optically and x-ray transparent. Each tool is mounted on a 0.025”/0.64 mm diameter non-magnetic stainless steel base. The base is fitted in a handle 0.7 mm diameter and included in each kit.