SupaMega Stainless Steel Base Molds, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62354-36 62354-60
102094-032PK 96.62 USD
102094-032 102094-034
SupaMega Stainless Steel Base Molds, Electron Microscopy Sciences
Moulds Moulds for Histology
The SupaMega range of stainless steel base molds have extra deep side walls allowing wax to be decanted into the cassettes during embedding without the risk of the usual leakage of wax.
The smaller faced mold also saves wax on medium sized blocks too large to fit into standard cassettes negating the use of full size molds.