Silicon carbide, abrasive film discs, Diameter 8" Plain Back

Supplier: Electron Microscopy Sciences

50257-1 50257-15 50257-3 50257-30 50257-5 50257-60 50257-9
102092-754PK 81.86 USD
102092-754 102092-756 102092-758 102092-760 102092-762 102092-764 102092-766
Silicon carbide, abrasive film discs, Diameter 8" Plain Back
Silicon carbide
Formula: SiC
MW: 40.1 g/mol
Melting Pt: 2700
MDL Number: MFCD00049531
CAS Number: 409-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR