Weigert's Resorcin Fuchsin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26370-05
26370-04
26370-03
26370-01
26370-02
101411-300EA
45.39
USD
101411-300
101411-298
101411-296
101411-292
101411-294
Weigert's Resorcin Fuchsin, Electron Microscopy Sciences
Stains
Histology Stains
For staining elastic fibers in blood vessel walls, nuclei and collagen.
- Elastic Tissues: Dark blue
- Nuclei (red if Orth's Carmine used): Dark blue
- Collagen: Red-Pink
- Miscellaneous Tissue Elements: Yellow (with Van Gieson's stain)
Certificaciones: Certificates of analysis are available.