Lillie Modification of Masson’s Trichrome, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26364-08
26364-01
26364-03
26364-04
26364-05
26364-06
26364-07
26364-7A
26364-02
101411-274EA
40.01
USD
101411-274
101411-260
101411-264
101411-266
101411-268
101411-270
101411-272
101411-276
101411-262
Lillie Modification of Masson’s Trichrome, Electron Microscopy Sciences
Stains
Histology Stains
Lillie Modification reagent of Masson’s Trichrome
Lillie (1940)
Stains cells, cytoplasm muscle and collagen of mammalian tissue