Alcojet® Low Foaming Powdered Detergents, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60513-01 60513-09
102093-282EA 105.34 USD
102093-282 102093-284
Alcojet® Low Foaming Powdered Detergents, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Non-ionic, low foaming cleaner for use in labware washers, parts washers, power spray systems, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. pH is 12.
  • Free rinsing for reliable results, no interfering residues
  • USDA authorized
  • Dilute 1:200