Technovit® 4000 Embedding Medium, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14656-03 14656-02
101410-650EA 709.73 USD
101410-650 101410-648
Technovit® 4000 Embedding Medium, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Technovit® 4000 is the low shrinkage embedding medium
It is a fast curing, cold polymerizing, three-component resin, which is based on a modified polyester. It is available in the form of powder, syrup I, and syrup II mixed at a ratio of 2:2:1 (powder:syrup I:syrup II). Syrup I and Syrup II are mixed together first and the powder is mixed in last. The color is white opaque. It shows an excellent margin fit and an optimum grinding and polishing properties.