W PC-620 Ultrasonic Cleaner, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71046-R2 71046-E1 71046-E2
100493-098EA 2011.67 USD
100493-098 100493-100 100493-094 100493-096
W PC-620 Ultrasonic Cleaner, Electron Microscopy Sciences
Ultrasonic Cleaners
The PC-620 is offered in 2 frequencies to match cleaning capability to your application. At 40 kHz it is ideal for cleaning pipettes, lab glassware, and other items requiring general cleaning.