Wehnelt Apertures, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
49100-TA 49050-TA 49040-TA 49030-TA 49020-TA 49075-TA
100496-784EA 282.02 USD
100496-784 100496-780 100496-778 100488-078 100488-076 100496-782
Wehnelt Apertures, Electron Microscopy Sciences
Microscopes
Made from 0.004" tantalum foil. Replaceable tantalum apertures for some microscopes (Amray and Philips). They improve the performance of the electron gun.