Paraplast Plus, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
19216 19217
102091-006EA 56.72 USD
102091-006 102091-008
Paraplast Plus, Electron Microscopy Science
Stains Microscopy and staining reagents
Reduces infiltration time by one-third. Cuts to 2 micron thickness with excellent ribbon continuity.
  • Convenient pellet form melts rapidly
  • Melting point 56°C
  • Double filtered
  • No need for filtration before use.