P47 Scintillators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
82010 82012 82017 82029 82014 82021 82022 82028 82030 82032 82016 82013 82026 82018
100492-396EA 168.69 USD
100492-396 100492-398 100492-406 100492-422 100492-402 100492-412 100492-414 100492-420 100492-424 100492-426 100492-404 100492-400 100492-418 100492-408
P47 Scintillators, Electron Microscopy Sciences
Lights
These scintillators are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required.