Oven Incubator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63230-10 63230-20
100498-358EA 2578.08 USD
100498-358 102094-554
Oven Incubator, Electron Microscopy Sciences
Incubators Standard Incubators
This Oven/Incubator with aluminum inner chamber and venting is up to the task of providing many years of long service in busy labs.
  • Oven works on the gravity convection heating method
  • An aluminum chamber provides excellent temperature uniformity
  • A thermometer port at the top of the oven accommodates any partial immersion thermometer
  • Heats up rapidly (56°C in 9 minutes)
  • The chamber is vented through the top to allow for the removal of moisture and other vapors
  • The unit comes complete with one removable metal shelf, three-wire cord, and plug
  • Use for general drying and incubation applications and polymerization of resins
The cast aluminum chamber provides for dependable temperature uniformity and the vent efficiently expels moisture and vapors. The unit comes complete with a removable shelf.
Overall Dimensions: HxWxD: 225 x185 x135 mm