SuperNuova™ Digital Hot Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72342-12 72342-20 72342-52 72342-60 72345-12 72345-20
102097-916EA 782.87 USD
102097-916 102097-918 102097-920 102097-922 102097-938 102097-940
SuperNuova™ Digital Hot Plates, Electron Microscopy Sciences
Heaters Hot Plates
Hot plates with rugged low profile cast aluminum body and ceramic or aluminum top plate.
  • Warning system deters accidental contact with hot surface
  • Remote probe allow for accurate reading of solution temperature to 0.1 °C for temperature below 100 °C
  • User-adjustable over-temperature protection prevents runaway temperature conditions
  • Rugged low profile cast aluminum body provides stability and durability
  • Unit designed provides protection of internal components in case of spillage
  • Available in 7 x 7" and 10 x 10" sizes
Features:
- Accommodates sampleMore Product Information