Veco Grids with Handle, Single Slot, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HD2010-NI HD2010-CU
100495-858EA 58.46 USD
100495-858 100495-856
Veco Grids with Handle, Single Slot, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics.
  • With a 0.8 mil thickness
  • Measures 3.05 mm in diameter
Veco Grids are the most rigid grids available. Material: Copper (Cu); Nickel (Ni).