Anti-DES Mouse Monoclonal Antibody [clone: D33]

Supplier: Diagnostic Biosystems

MOB060 PDM006 MOB060-05
99990-232EA 564.25 USD
99990-232 99990-826 95024-512
Anti-DES Mouse Monoclonal Antibody [clone: D33]
Antibodies
This antibody reacts with desmin protein of 53 kDa. Both striated as well as smooth muscle cells stain well with this antibody. The antibody shows a broad interspecific cross-reactivity, reacting with human.

Type:
Antigen: DES
Clonality: Monoclonal
Clone: D33
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR