Anti-CK8/18 Mouse Monoclonal Antibody [clone: 5D3]

Supplier: Diagnostic Biosystems

MOB189 PDM070 MOB189-05
99990-396EA 502.01 USD
99990-396 99990-924 95024-462
Anti-CK8/18 Mouse Monoclonal Antibody [clone: 5D3]
Antibodies
This antibody is specific to human cytokeratins 8 (52.5 kDa) and 18 (45 kDa). This antibody stains simple and glandular epithelium. This antibody can be useful for the identification of adenocarcinomas and most squamous cell carcinomas.

Type:
Antigen: CK8/18
Clonality: Monoclonal
Clone: 5D3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR