Anti-CK8 Mouse Monoclonal Antibody [clone: 35[beta]E11]

Supplier: Diagnostic Biosystems

PDM117 MOB054-05 MOB054
99991-008EA 247.7 USD
99991-008 95024-460 99990-220
Anti-CK8 Mouse Monoclonal Antibody [clone: 35[beta]E11]
Antibodies
This antibody recognizes keratin polypeptide of 54 kDa. It stains strongly with non-squamous epithelium but does not stain squamous epithelium. It reacts with the sweat gland and ducts in the skin. Adenocarcinomas of the ovary, gastrointestinal tract, ductal carcinoma of breast, pancreas and bile ducts, stain positive with this antibody.

Type:
Antigen: CK8
Clonality: Monoclonal
Clone: 35[beta]E11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR