Anti-CK5 Mouse Monoclonal Antibody [clone: XM26]

Supplier: Diagnostic Biosystems

MOB361-05 PDM139 MOB361
95024-446EA 443.73 USD
95024-446 95029-752 99990-674
Anti-CK5 Mouse Monoclonal Antibody [clone: XM26]
Antibodies
This antibody reacts with cytokeratin 5 intermediate filament protein. Basal cells of human epidermis express acidic cytokeratin 14 and basic cytokeratin 5. Cytokeratin 5 is a 58 kDa protein, which is closely related to cytokeratin 6. Cytokeratin 5 and 6 are expressed in basal cell epithelioma, basal cells of prostate, urothelium, vagina, squamous cell carcinomas of skin, tongue, epiglottis and rectal-anal region. Cytokeratin 5 can be useful in the distinction of mesotheliomas from most adenocarcinomas.

Type:
Antigen: CK5
Clonality: Monoclonal
Clone: XM26
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen Symbol CK5 CK5
Environmentally Preferable
Antigen Name Cytokeratin 5 Cytokeratin 5
Antigen Synonyms CK-5|58 kda cytokeratin|cytokeratin-5|type-II keratin Kb5|Dowling-Meara/Kobner/Weber-Cockayne types)|type II cytoskeletal 5|epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types|keratin 5 (epidermolysis bullosa simplex|keratin CK-5|58 kda cytokeratin|cytokeratin-5|type-II keratin Kb5|Dowling-Meara/Kobner/Weber-Cockayne types)|type II cytoskeletal 5|epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types|keratin 5 (epidermolysis bullosa simplex|keratin
Clonality Monoclonal Monoclonal
Clone XM26 XM26
Reactivity
Host Mouse Mouse
Isotype IgG1 kappa IgG1 kappa
Concentration Concentrated Ready to use

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR