Johns Hopkins Bulldog Clamps, Roboz Surgical
Supplier: Roboz Surgical
RS-7445 RS-7443 RS-7441 RS-7444 RS-7440 RS-7446 RS-7442 RS-7447
100188-872EA 130.15 USD
100188-872 100188-876 100188-908 100188-870 100188-906 100188-918 100188-890 100188-920
Johns Hopkins Bulldog Clamps, Roboz Surgical
Clamps General Purpose Clamps
The Bulldog clamp is a useful instrument for stopping blood flow to a particular organ of interest
These premium quality bulldog clamps have a rivet in the shank. The jaws can be separated by twisting the shanks sideways around the rivet. This provides easy access to the serrations in the jaws for thorough cleaning, which is very difficult when the jaws don't open wide enough or when the closing force keeps them shut during the cleaning cycle.
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.