Anti-CD8 Rabbit Monoclonal Antibody [clone: SP16]

Supplier: Diagnostic Biosystems

RMAB012 RMPD012
99991-150EA 1197.37 USD
99991-150 95025-180
Anti-CD8 Rabbit Monoclonal Antibody [clone: SP16]
Antibodies
This antibody recognizes the CD8. The CD8 molecule consists of two chains, α and β, which are expressed as a disulphide-linked α/β heterodimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not β chain).

Type:
Antigen: CD8
Clonality: Monoclonal
Clone: SP16
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR