Anti-CD23 Mouse Monoclonal Antibody [clone: 1B12]

Supplier: Diagnostic Biosystems

MOB294-05 PDM143 MOB294
95024-328EA 604 USD
95024-328 95029-760 99990-554
Anti-CD23 Mouse Monoclonal Antibody [clone: 1B12]
Antibodies
This antibody is specific to a membrane glycoprotein of 45 kDa, designated as CD23. CD23 is found on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocytic leukemias.

Type:
Antigen: CD23
Clonality: Monoclonal
Clone: 1B12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR