Anti-CASP3 0 Polyclonal Antibody

Supplier: Diagnostic Biosystems

RP096-05 RP096
95024-926EA 537.76 USD
95024-926 99991-364
Anti-CASP3 0 Polyclonal Antibody
Antibodies
This antibody reacts with a 32 kDa protein. Caspase 3 is a member of the interleukin-1 β-converting enzyme family. Caspase 3 is thought to be associated with induction of apoptosis. Caspase 3 is synthesized as inactive 32 kDa proenzyme and is processed during apoptosis generating two subunits of 17 kDa and 12 kDa. Caspase 3 stains the epithelial cells of skin, renal proximal tubules and collecting ducts.

Type:
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: 0
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR