Anti-CA 19-9 Mouse Monoclonal Antibody [clone: C241:5:1:4]

Supplier: Diagnostic Biosystems

MOB109 MOB109-05
99990-302EA 566.89 USD
99990-302 95024-256
Anti-CA 19-9 Mouse Monoclonal Antibody [clone: C241:5:1:4]
Antibodies
This antibody reacts specifically with sialyl Lewisa and recognizes an epitope being designated CA 19-9. It does not cross-reacts with Lewisa and Lewisb. CA 19-9 has been shown to be useful marker in the diagnosis and management of gastrointestinal cancers.

Type:
Antigen: CA 19-9
Clonality: Monoclonal
Clone: C241:5:1:4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR